Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper, Granular, 20 to 30 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7440-50-8
Cu
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 8.94kg
Quantity 2.5kg
Mesh Size 20 to 30 Mesh
SMILES [Cu]
IUPAC Name copper
Packaging Poly Bottle
Formula Weight 63.55
Physical Form Granular
Chemical Name or Material Copper
Grade BAKER ANALYZED™ Reagent