Alfa Aesar™ Copper(I) bromide-dimethyl sulfide complex

Manufacturer: Alfa Aesar™ 1936804

Quantity 2g

 View more versions of this product

Catalog No. AA1936804

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 54678-23-8
    Formula Weight 205.58
    MDL Number MFCD00043295
    UN Number UN3335
    Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex

    Specifications

    Formula Weight 205.58
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Physical Form Crystalline
    MDL Number MFCD00043295
    CAS 54678-23-8
    Melting Point 124° to 129°C (dec.)
    Quantity 2g
    Health Hazard 1 Warning
    Sensitivity Air & Moisture Sensitive
    UN Number UN3335
    Color Pale brown or pale green
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
    Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit