Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) chloride, 97%, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
2kg
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
Assay Percent Range 97%
Appearance Powder
Solubility Information Very soluble in concentrated HCl. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water.
Formula Weight 98.99
Physical Form Powder
Density 4.14
Refractive Index 1.93
Sensitivity Air and moisture sensitive
Boiling Point 1490°C
Melting Point 430°C
Quantity 2kg
Chemical Name or Material Copper(I) chloride
Show More Show Less

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H318

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-842-9

RTECSNumber : GL6990000

TSCA : Yes

storageNote1 : Air & Moisture Sensitive

Recommended Storage : Ambient temperatures