Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper(I) chloride, 99%, extra pure, purified, ACROS Organics™
Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
Specifications
Assay Percent Range | 98.5 to 101.5% (Complexometry) |
Linear Formula | CuCl |
Fieser | 01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97 |
Merck Index | 15,265 |
Solubility Information | Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh,forming complexes,practically insoluble in alcohol and acetone |
Formula Weight | 99 |
Physical Form | Crystalline Powder |
Percent Purity | 99% |
Grade | Extra Pure |
Density | 4.1400g/mL |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 231-842-9
RTECSNumber : GL6990000
TSCA : TSCA