Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) chloride, 99%, extra pure, purified, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
7758-89-6
ClCu
99
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
7758-89-6
ClCu
99
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula CuCl
Fieser 01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97
Merck Index 15,265
Solubility Information Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh,forming complexes,practically insoluble in alcohol and acetone
Formula Weight 99
Physical Form Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Density 4.1400g/mL
Iron (Fe) 200ppm max.
Packaging Glass bottle
Solubility Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone
Specific Gravity 4.14
Sulfate 500ppm max.
Boiling Point 1490.0°C
Color Gray-Green
Melting Point 430.0°C
Quantity 500g
Chemical Name or Material Copper(I) chloride, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 231-842-9

RTECSNumber : GL6990000

TSCA : TSCA