Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Copper(I) Chloride, ≥99%, ReagentPlus™, Purified, Honeywell™

Click to view available options
Quantity:
25g
100g
Packaging:
Poly Bottle
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride,copper i chloride,copper chloride cucl,copper monochloride,dicopper dichloride,copper 1+ chloride,cucl,chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]

Reagent Grade, purified,≥99%

  • ≥95% for general laboratory use.
Linear Formula CuCl
UN Number UN2802
Appearance Powder
Formula Weight 99g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Packaging Poly Bottle
Vapor Pressure 1.3mmHg (546°C)
Boiling Point 1490°C
Color Gray
Melting Point 430°C
Quantity 25g
Chemical Name or Material Copper(I) Chloride, Purified
Show More Show Less

P273-P280-P305 + P351 + P338-P501

H302-H315-H318-H410

DOTInformation : 8/PGIII

EINECSNumber : 231-842-9

RTECSNumber : GL6990000