Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (I) Chloride, Anhydrous
SCI_ED

Catalog Number p-4416376
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade - Powder
Reagent Grade - Powder
This item is not returnable. View return policy
7758-89-6
ClCu
98.996
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
This item is not returnable. View return policy
7758-89-6
ClCu
98.996
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]

This product is for educational use - no C of A is available.

Color Gray to Green
Physical Form Solid
Quantity 100 g
Linear Formula CuCl1
Formula Weight 99
Packaging Lab Grade - Powder
Density 4.14g/mL
Chemical Name or Material Copper (I) Chloride, Anhydrous
ShelfLife 2 years
DOTInformation DOT Class 8, III, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit