Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) iodide, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
7681-65-4
CuI
190.45
MFCD00010978
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
7681-65-4
CuI
190.45
MFCD00010978
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
C-X bond formation reagent
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula CuI
Fieser 01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88
Merck Index 15, 2652
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol
Formula Weight 190.45
Physical Form Powder
Percent Purity 98%
Iron (Fe) 50ppm max.
Packaging Glass bottle
Sulfate 5000ppm max.
Boiling Point 1290.0°C
Color Brown-Gray to White
Melting Point 605.0°C
Quantity 250g
Chemical Name or Material Copper(I) iodide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 231-674-6