Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) oxide, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Glass bottle
1317-39-1
Cu2O
143.09
MFCD00010974
Cuprous oxide
1317-39-1
Cu2O
143.09
MFCD00010974
Cuprous oxide
Assay Percent Range 96% min. (Complexometry)
Linear Formula Cu2O
Merck Index 15, 2654
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia or min.acids
Formula Weight 143.09
Physical Form Powder
Percent Purity 97%
Iron (Fe) 500ppm max.
Nickel (Ni) 100ppm max.
Packaging Glass bottle
Silver (Ag) 50ppm max.
Solubility Solubility in water: practically insoluble. Other solubilities: soluble in ammonia or min.acids
Boiling Point 1800.0°C
Color Brown-Red
Melting Point 1232.0°C
Quantity 25g
Chemical Name or Material Copper(I) oxide, 97%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 215-270-7

RTECSNumber : GL8050000

TSCA : TSCA