Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper(I) oxide, 99.9% (metals basis), Alfa Aesar™
Click to view available options
Quantity:
5g
25g
100g
500g
Specifications
Assay Percent Range | 99% (Metal basis) |
Appearance | Purple-red powder |
Solubility Information | Insoluble in water but soluble in acids. |
Formula Weight | 143.08 |
Physical Form | Powder |
Density | 6 |
Odor | Odorless |
Refractive Index | 2.705 |
Sensitivity | Air and moisture sensitive |
Boiling Point | 1800°C (decomposition) |
Show More |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
H302+H332-H318
DOTInformation : DOT Class: 9, Packing Group: III
EINECSNumber : 215-270-7
RTECSNumber : GL8050000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen