Learn More
Copper(I) oxide, 99% (metals basis), Thermo Scientific Chemicals
Description
Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1800°C (decomposition) |
Melting Point | 1235°C |
Color | Purple-Red |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99% (Metal basis) |
Solubility Information | It is insoluble in water and organic solvents. |
Formula Weight | 143.08 |
Odor | Odorless |
Refractive Index | 2.705 |
Show More |
Safety and Handling
Hazard Category | H302+H332-H318 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c |
DOTInformation | DOT Class: 9, Packing Group: III |
EINECSNumber | 215-270-7 |
RTECSNumber | GL8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.