Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407431
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
1317-39-1
Cu2O
MFCD00010974
1317-39-1
Cu2O
MFCD00010974

Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Color Purple-Red
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metal basis)
Solubility Information It is insoluble in water and organic solvents.
Formula Weight 143.08
Odor Odorless
Refractive Index 2.705
Sensitivity Air and moisture sensitive
Appearance Purple-red powder
Density 6
Chemical Name or Material Copper(I) oxide
Show More Show Less
Hazard Category H302+H332-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
DOTInformation DOT Class: 9, Packing Group: III
EINECSNumber 215-270-7
RTECSNumber GL8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit