Alfa Aesar™ Copper(I) selenide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 4566018

Quantity 50g

 View more versions of this product

Catalog No. AA4566018

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 20405-64-5
    Molecular Formula Cu2Se
    Formula Weight 206.05
    MDL Number MFCD00049908
    UN Number UN3077
    Merck Index 14,2666
    Chemical Name or Material Copper (I) selenide

    Specifications

    Formula Weight 206.05
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Pieces
    CAS 20405-64-5
    Health Hazard 1 Danger
    Density 6.84g/mL
    TSCA TSCA
    Particle Size 2mm & down
    Color Dark blue to black
    Quantity 50g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Molecular Formula Cu2Se
    MDL Number MFCD00049908
    UN Number UN3077
    Merck Index 14,2666
    Melting Point 1113°C
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Copper (I) selenide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit