Alfa Aesar™ Copper(I) trifluoromethanesulfonate benzene complex (2:1)

Manufacturer: Alfa Aesar™ 1698614

Quantity 25g

 View more versions of this product

Catalog No. AA1698614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37234-97-2
    Formula Weight 503.33
    MDL Number MFCD00043300
    UN Number UN3181
    Synonym Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
    Chemical Name or Material Copper(I) trifluoromethanesulfonate benzene complex (2:1)

    Specifications

    Formula Weight 503.33
    Health Hazard 3 GHS P Statement
    P210-P260-P280-P240-P314-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3181
    Health Hazard 2 GHS H Statement
    H372-H228
    Causes damage to organs through prolonged or repeated exposure.
    Flammable solid.
    Physical Form Powder
    MDL Number MFCD00043300
    CAS 37234-97-2
    Quantity 25g
    Synonym Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
    Solubility Information Soluble in hot benzene
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Chemical Name or Material Copper(I) trifluoromethanesulfonate benzene complex (2:1)

    GHS H Statement
    H372-H228
    Causes damage to organs through prolonged or repeated exposure.
    Flammable solid.

    GHS P Statement
    P210-P260-P280-P240-P314-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit