Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(I) trifluoromethanesulfonate benzene complex (2:1)

Manufacturer:  Alfa Aesar™ 1698603

Quantity 1g

 View more versions of this product

Catalog No. AA1698603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

37234-97-2
MFCD00043300
Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
503.33
UN3181
Copper(I) trifluoromethanesulfonate benzene complex (2:1)

Specifications

37234-97-2
Air & Moisture Sensitive
503.33
UN3181
Soluble in hot benzene
1g
Powder
MFCD00043300
Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
Copper(I) trifluoromethanesulfonate benzene complex (2:1)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H372-H228
Causes damage to organs through prolonged or repeated exposure.
Flammable solid.

GHS P Statement
P210-P260-P280-P240-P314-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit