Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper ICP Standard, 1mL = 10mg Cu (10,000ppm Cu), Ricca Chemical

Click to view available options
Quantity:
50mL
100mL
500mL
Packaging:
Poly natural
7440-50-8,7697-37-2,7732-18-5
Cu
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052
copper
[Cu]
7440-50-8,7697-37-2,7732-18-5
Cu
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052
copper
[Cu]
CAS Min % 88.0,8.94,0.88
CAS Max % 92.0,9.31,0.92
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 10,000ppm Cu
Equivalence 1 mL = 10 mg Cu
Color Blue
Quantity 100mL
Chemical Name or Material Copper ICP Standard