Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) 2,9,16,23-Tetra-tert-butylphthalocyanine 97.0+%, TCI America™

Click to view available options
Quantity:
100mg
39001-64-4
C48H48CuN8
800.514
MFCD00059117
IDACJLJAKMQWEW-UHFFFAOYSA-N
4,4′C,4′C′C,4′C′C′C-Tetra-tert-butylphthalocyanine Copper(II), 2,9,16,23-Tetra-tert-butylphthalocyanine Copper(II)
15325151
CC(C)(C)C1=CC2=C(C=C1)C3=NC4=NC(=NC5=C6C=C(C=CC6=C([N-]5)N=C7C8=C(C=CC(=C8)C(C)(C)C)C(=N7)N=C2[N-]3)C(C)(C)C)C9=C4C=C(C=C9)C(C)(C)C.[Cu+2]
39001-64-4
C48H48CuN8
800.514
MFCD00059117
IDACJLJAKMQWEW-UHFFFAOYSA-N
4,4′C,4′C′C,4′C′C′C-Tetra-tert-butylphthalocyanine Copper(II), 2,9,16,23-Tetra-tert-butylphthalocyanine Copper(II)
15325151
CC(C)(C)C1=CC2=C(C=C1)C3=NC4=NC(=NC5=C6C=C(C=CC6=C([N-]5)N=C7C8=C(C=CC(=C8)C(C)(C)C)C(=N7)N=C2[N-]3)C(C)(C)C)C9=C4C=C(C=C9)C(C)(C)C.[Cu+2]
Color Blue-Purple
Quantity 100mg
Formula Weight 800.51
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material Copper(II) 2,9,16,23-Tetra-tert-butylphthalocyanine