Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) acetate monohydrate, 98+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
6046-93-1
C4H6CuO4·H2O
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate,cupric acetate monohydrate,copper diacetate monohydrate,copper acetate monohydrate,copper 2+ acetate, monohydrate,acetic acid, copper 2+ salt, monohydrate,verdigris,copper acetate hydrate,diacetoxycopper hydrate,copper acetate, hydrate
165397
copper;diacetate;hydrate
CC(=O)[O-].CC(=O)[O-].O.[Cu+2]
6046-93-1
C4H6CuO4·H2O
199.65
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate,cupric acetate monohydrate,copper diacetate monohydrate,copper acetate monohydrate,copper 2+ acetate, monohydrate,acetic acid, copper 2+ salt, monohydrate,verdigris,copper acetate hydrate,diacetoxycopper hydrate,copper acetate, hydrate
165397
copper;diacetate;hydrate
CC(=O)[O-].CC(=O)[O-].O.[Cu+2]
Oxidation reagent
Assay Percent Range 98% min. (Iodometry)
Linear Formula (CH3CO2)2Cu·H2O
Merck Index 15,2614
Solubility Information Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 199.65
Physical Form Crystalline Powder
Percent Purity 98+%
Grade Extra Pure
Chloride 30ppm max
Chlorine (Cl) 30ppm max.
Density 1.88g/mL
Infrared Spectrum Authentic
Insoluble Matter 0.01% max
Iron (Fe) 20ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
Sulfate 100ppm max.
Boiling Point 240.0°C
Color Blue to Blue-Green
Melting Point 115.0°C
Quantity 500g
Chemical Name or Material Copper(II) acetate monohydrate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger