Alfa Aesar™ Copper(II) acetate monohydrate, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3548122

Quantity 100g

 View more versions of this product

Catalog No. AA3548122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6046-93-1
    Linear Formula Cu(OOCCH3)2·H2O
    Formula Weight 199.65 (181.64 Anhydrous)
    MDL Number MFCD00149570
    UN Number UN3077
    Merck Index 14,2624
    Chemical Name or Material Copper(II) acetate monohydrate

    Specifications

    Formula Weight 199.65 (181.64 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Mesh Size −4 Mesh
    Odor Odorless
    Density 1.882g/mL
    UN Number UN3077
    Color Blue-green
    Melting Point 115°C
    pH 5.2 to 5.5
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Linear Formula Cu(OOCCH3)2·H2O
    MDL Number MFCD00149570
    CAS 6046-93-1
    Merck Index 14,2624
    Quantity 100g
    RTECS Number AG3500000
    Chemical Name or Material Copper(II) acetate monohydrate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit