Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper(II) chloride, anhydrous, 99%, extra pure, ACROS Organics™
Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
Description
C-X bond formation reagent
Specifications
Additional Information | cupric nitrate: < 0.75% |
Assay Percent Range | 98.5% min. (CuCl2) |
Linear Formula | CuCl2 |
UN Number | UN 2802 |
Fieser | 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 10,106; 11,267; 13,152; 14,100 |
Merck Index | 15, 2623 |
Solubility Information | Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol,acetone and ether |
Formula Weight | 134.45 |
Physical Form | Fine Crystalline Powder |
Percent Purity | 99% |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical
GHS Signal Word: Danger
EINECSNumber : 231-210-2