Alfa Aesar™ Copper(II) ethoxide

Manufacturer: Alfa Aesar™ 1886206

Quantity 5g

 View more versions of this product

Catalog No. AA1886206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2850-65-9
    Linear Formula Cu(OC2H5)2
    Formula Weight 153.67
    MDL Number MFCD00015650
    UN Number UN3181
    Chemical Name or Material Copper(II) ethoxide

    Specifications

    Formula Weight 153.67
    Health Hazard 3 GHS P Statement
    P210-P280h-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H228-H302-H315-H319-H335
    Flammable solid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Green
    Quantity 5g
    Sensitivity Moisture Sensitive
    MDL Number MFCD00015650
    UN Number UN3181
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Linear Formula Cu(OC2H5)2
    Health Hazard 1 Warning
    Melting Point ≈120°C (dec.)
    CAS 2850-65-9
    Odor Alcohol-like
    Chemical Name or Material Copper(II) ethoxide

    GHS H Statement
    H228-H302-H315-H319-H335
    Flammable solid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P280h-P305+P351+P338-P337+P313
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit