Alfa Aesar™ Copper(II) ethoxide, 98%

Manufacturer: Alfa Aesar™ 4631606

Quantity 5g

 View more versions of this product

Catalog No. AA4631606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2850-65-9
    Molecular Formula C4H10CuO2
    Linear Formula Cu(OC2H5)2
    Formula Weight 153.67
    MDL Number MFCD00015650
    UN Number UN3181
    Chemical Name or Material Copper (II) ethoxide

    Specifications

    Formula Weight 153.67
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈120°C (dec.)
    Linear Formula Cu(OC2H5)2
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Green
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00015650
    CAS 2850-65-9
    UN Number UN3181
    Health Hazard 1 Warning
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Molecular Formula C4H10CuO2
    Quantity 5g
    Chemical Name or Material Copper (II) ethoxide
    Assay 98%

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit