Alfa Aesar™ Copper(II) ethylacetoacetate, 97%

Manufacturer: Alfa Aesar™ 4148930

Quantity 250g

 View more versions of this product

Catalog No. AA4148930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14284-06-1
    Formula Weight 321.81
    MDL Number MFCD00017759
    Chemical Name or Material Copper(II) ethylacetoacetate

    Specifications

    Formula Weight 321.81
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Physical Form Powder
    MDL Number MFCD00017759
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 192°C
    CAS 14284-06-1
    Quantity 250g
    Solubility Information Soluble in ethanol
    Health Hazard 1 Warning
    Odor Odorless
    TSCA TSCA
    Chemical Name or Material Copper(II) ethylacetoacetate
    Assay 97%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit