Alfa Aesar™ Copper(II) fluoride hydrate, Puratronic™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1300109

Quantity 10g

 View more versions of this product

Catalog No. AA1300109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13454-88-1
    Formula Weight 101.54 (Anhydrous)
    MDL Number MFCD00010977
    UN Number UN3288
    Merck Index 14,2638
    Chemical Name or Material Copper(II) fluoride hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 101.54 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.
    UN Number UN3288
    Merck Index 14,2638
    Color Blue
    Melting Point >130°C (dec.)
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    Quantity 10g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    MDL Number MFCD00010977
    CAS 13454-88-1
    Density 2.934g/mL
    Chemical Name or Material Copper(II) fluoride hydrate
    Assay 99.99%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.

    GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit