Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(II) formate tetrahydrate, 98%

Click to view available options
Quantity:
50g
250g
5893-61-8
C2H10CuO8
225.64
MFCD00167128
LSIWWRSSSOYIMS-UHFFFAOYSA-L
copper ii formate tetrahydrate,cupric formate tetrahydrate,copper diformate tetrahydrate,unii-v7ra28204j,copper 2+ formate tetrahydrate,cupric formate tetrahydrate mi,copper formate cu hcoo 2 tetrahydrate,formic acid copper salt 2:1 tetrahydrate,acmc-209m8c
6432244
copper;diformate;tetrahydrate
C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2]
5893-61-8
C2H10CuO8
225.64
MFCD00167128
LSIWWRSSSOYIMS-UHFFFAOYSA-L
copper ii formate tetrahydrate,cupric formate tetrahydrate,copper diformate tetrahydrate,unii-v7ra28204j,copper 2+ formate tetrahydrate,cupric formate tetrahydrate mi,copper formate cu hcoo 2 tetrahydrate,formic acid copper salt 2:1 tetrahydrate,acmc-209m8c
6432244
copper;diformate;tetrahydrate
C(=O)[O-].C(=O)[O-].O.O.O.O.[Cu+2]
Applications
It is employed as an antibacterial agent in the treatment of cellulose.

Solubility
Soluble in water (125 g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 50g
UN Number UN3077
Merck Index 14,2639
Solubility Information Soluble in water (125g/L at 20°C). Very slightly soluble in alcohol. Practically insoluble in most organic solvents.
Formula Weight 225.65 (153.59 Anhydrous)
Percent Purity 98%
Chemical Name or Material Copper(II) formate tetrahydrate

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 208-865-8

TSCA : Yes

Recommended Storage : Ambient temperatures