Alfa Aesar™ Copper(II) gluconate

Manufacturer: Alfa Aesar™ 15325A1

Quantity 1kg

 View more versions of this product

Catalog No. AA15325A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 527-09-3
    Formula Weight 453.85
    MDL Number MFCD00075297
    Merck Index 14,2640
    Chemical Name or Material Copper(II) gluconate

    Specifications

    Formula Weight 453.85
    MDL Number MFCD00075297
    CAS 527-09-3
    Physical Form Powder
    Quantity 1kg
    TSCA TSCA
    Melting Point 155° to 157°C
    Merck Index 14,2640
    RTECS Number LZ5058000
    Chemical Name or Material Copper(II) gluconate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit