Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(II) hexafluoro-2,4-pentanedionate hydrate, 98%

Click to view available options
Quantity:
1g
5g
25g
155640-85-0
C10H6CuF12O5
497.68
MFCD00151019
DBDCQCFMFYPMNY-WOGKQDBSSA-N
copper ii hexafluoroacetylacetonate hydrate,bis hexafluoroacetylacetonato copper ii hydrate,copper ii hexafluoro-2,4-pentanedionate hydrate,copper hexafluoroacetylacetonate hydrate,copper ii hexafluoroacetyl-acetonate hydrate,hexafluoroacetylacetonato copper ii hydrate,copper ii hexafluoroacetylacetonate hydrate, elec. gr.,bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate,copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate,3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate
56845342
copper;(E)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.O.[Cu]
155640-85-0
C10H6CuF12O5
497.68
MFCD00151019
DBDCQCFMFYPMNY-WOGKQDBSSA-N
copper ii hexafluoroacetylacetonate hydrate,bis hexafluoroacetylacetonato copper ii hydrate,copper ii hexafluoro-2,4-pentanedionate hydrate,copper hexafluoroacetylacetonate hydrate,copper ii hexafluoroacetyl-acetonate hydrate,hexafluoroacetylacetonato copper ii hydrate,copper ii hexafluoroacetylacetonate hydrate, elec. gr.,bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate,copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate,3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate
56845342
copper;(E)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.C(=C(C(F)(F)F)O)C(=O)C(F)(F)F.O.[Cu]
Formula Weight 477.64 (Anhydrous)
Percent Purity 98%
Boiling Point 220°C
Melting Point ~100°C
Quantity 25g
Chemical Name or Material Copper(II) hexafluoro-2,4-pentanedionate hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning