Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(II) methoxide, typically 98% (metals basis)

Manufacturer:  Alfa Aesar™ 1886103

Quantity 1g

 View more versions of this product

Catalog No. AA1886103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1184-54-9
125.61
UN3181
Cu(OCH3)2
MFCD00015611
Copper(II) methoxide

Specifications

≈206°C (dec.)
1g
Green
Powder
125.61
UN3181
98% (typically), (metals basis)
1184-54-9
Alcohol-like
Moisture Sensitive
Cu(OCH3)2
MFCD00015611
Copper(II) methoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280h-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit