Copper (II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Alfa Aesar

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths

Manufacturer: Alfa Aesar 12523A9

Quantity 10kg

 View more versions of this product

Catalog No. AA12523A9

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19004-19-4
    Physical Form Crystalline Lump
    Assay Percent Range 98.0-102.0%
    Molecular Formula CuN2O6·2.5H2O
    Formula Weight 232.59 (187.55anhydrous)
    MDL Number MFCD00149671
    Synonym Cupric nitrate hemi(pentahydrate)
    Chemical Name or Material Copper(II) nitrate hemi(pentahydrate), trihydrate
    Grade ACS Grade

    Specifications

    Physical Form Crystalline Lump
    Formula Weight 232.59 (187.55anhydrous)
    Quantity 10kg
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 221-838-5
    Synonym Cupric nitrate hemi(pentahydrate)
    Assay Percent Range 98.0-102.0%
    Molecular Formula CuN2O6·2.5H2O
    MDL Number MFCD00149671
    CAS 19004-19-4
    Health Hazard 1 H272-H314-H318-H302; May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.
    Storage Note 1 Hygroscopic
    Chemical Name or Material Copper(II) nitrate hemi(pentahydrate), trihydrate
    Grade ACS Grade

    Hygroscopic

    H272-H314-H318-H302; May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit