Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) nitrate trihydrate, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
10031-43-3
CuN2O6·3H2O
241.6
MFCD00149671
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate,copper nitrate trihydrate,cupric nitrate trihydrate,copper ii nitrate hydrate,copper ii nitrate 3-hydrate,copper 2+ trihydrate dinitronate,copper 2+ trihydrate dinitrate,copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
10031-43-3
CuN2O6·3H2O
241.6
MFCD00149671
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate,copper nitrate trihydrate,cupric nitrate trihydrate,copper ii nitrate hydrate,copper ii nitrate 3-hydrate,copper 2+ trihydrate dinitronate,copper 2+ trihydrate dinitrate,copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
Assay Percent Range 98.5 to 104% (Iodometry)
Linear Formula Cu(NO3)2·3H2O
Fieser 17,366
Merck Index 15, 2633
Solubility Information Solubility in water: 267g/100 mL. Other solubilities: freely soluble in alcohol,practically insoluble in ethyl acetate
Formula Weight 241.6
Physical Form Fine Powder
Percent Purity 99%
Grade Analysis
Calcium (Ca) 50ppm max.
Chloride 20ppm max.
Chlorine (Cl) 20ppm max.
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate
Sulfate 50ppm max.
Trace Analysis 50ppm max.
Zinc (Zn) 50ppm max.
Color Brown to Cream
Melting Point 114.0°C
Quantity 2.5kg
Chemical Name or Material Copper(II) nitrate trihydrate, p.a.
Show More Show Less

GHS H Statement
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

RTECSNumber : GL7875000