Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) nitrate trihydrate, 99%, for analysis, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
10031-43-3
CuH6N2O9
241.599
MFCD00149671
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate,copper nitrate trihydrate,cupric nitrate trihydrate,copper ii nitrate hydrate,copper ii nitrate 3-hydrate,copper 2+ trihydrate dinitronate,copper 2+ trihydrate dinitrate,copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
10031-43-3
CuH6N2O9
241.599
MFCD00149671
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper ii nitrate trihydrate,copper nitrate trihydrate,cupric nitrate trihydrate,copper ii nitrate hydrate,copper ii nitrate 3-hydrate,copper 2+ trihydrate dinitronate,copper 2+ trihydrate dinitrate,copper ii nitrate trihydrate-cu puratrem
9837674
copper;dinitrate;trihydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
Assay Percent Range 99%
Linear Formula Cu(NO3)2·3H2O
Fieser 17,366
Merck Index 15, 2633
Formula Weight 241.6
Physical Form Fine Powder
Percent Purity 98.5 to 104.0% (Iodometry)
Grade Analysis
Calcium (Ca) 50ppm max.
Chloride 20ppm max.
Chlorine (Cl) 20ppm max.
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Glass Bottle
Sulfate 50ppm max.
Trace Analysis 50ppm max.
Zinc (Zn) 50ppm max.
Color Brown to Cream
Quantity 5g
Chemical Name or Material Copper(II) nitrate trihydrate, p.a.
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May intensify fire; oxidizer.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment.

Danger

RTECSNumber : GL7875000