Copper(II) oxide, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar 12299A3

Quantity 2kg

 View more versions of this product

Catalog No. AA12299A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-38-0
    Physical Form Powder
    Assay Percent Range 97%
    Molecular Formula CuO
    Formula Weight 79.55
    MDL Number MFCD00010979
    Chemical Name or Material Copper(II) oxide

    Specifications

    Formula Weight 79.55
    Quantity 2kg
    Color Black
    DOT Information DOT Class: 9, Packaging Group: III
    EINECS Number 215-269-1
    Physical Form Powder
    Assay Percent Range 97%
    Molecular Formula CuO
    Appearance Black powder
    MDL Number MFCD00010979
    CAS 1317-38-0
    Health Hazard 1 H400-H410-H302; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.
    Chemical Name or Material Copper(II) oxide

    H400-H410-H302; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit