Alfa Aesar™ Copper(II) oxide, ACS

Manufacturer: Alfa Aesar™ 1315236

Quantity 500g

 View more versions of this product

Catalog No. AA1315236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-38-0
    Formula Weight 79.55
    MDL Number MFCD00010979
    UN Number UN3077
    Merck Index 14,2646
    Chemical Name or Material Copper(II) oxide
    Grade ACS Grade

    Specifications

    Formula Weight 79.55
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3077
    Color Brown to black
    RTECS Number GL7900000
    Odor Odorless
    Physical Form Wire, Random Pieces
    Refractive Index 2.63
    Merck Index 14,2646
    Density 6.3g/mL
    CAS 1317-38-0
    Quantity 500g
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    MDL Number MFCD00010979
    Melting Point 1326°C
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material Copper(II) oxide
    Grade ACS Grade

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit