Alfa Aesar™ Copper(II) oxide, nanopowder

Manufacturer: Alfa Aesar™ 4466309

Quantity 10g

 View more versions of this product

Catalog No. AA4466309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-38-0
    Molecular Formula CuO
    Formula Weight 79.55
    MDL Number MFCD00010979
    UN Number UN3077
    Merck Index 14,2646
    Chemical Name or Material Copper (II) oxide

    Specifications

    Formula Weight 79.55
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Odorless
    Refractive Index 2.63
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number GL7900000
    Physical Form Nanopowder
    Molecular Formula CuO
    Surface Area 13m2/g
    Health Hazard 1 Warning
    Density 6.3g/mL
    MDL Number MFCD00010979
    CAS 1317-38-0
    UN Number UN3077
    Melting Point 1326°C
    Particle Size 30 to 50nm APS
    Merck Index 14,2646
    Color Black
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material Copper (II) oxide

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit