Alfa Aesar™ Copper(II) perchlorate hexahydrate, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4431909

Quantity 10g

 View more versions of this product

Catalog No. AA4431909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10294-46-9
    Molecular Formula Cu(ClO4)2·6H2O
    Formula Weight 370.53 (262.44 Anhydrous)
    MDL Number MFCD00149666
    UN Number UN1481
    Merck Index 14,2647
    Chemical Name or Material Copper (II) perchlorate hexahydrate

    Specifications

    Formula Weight 370.53 (262.44 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Sensitivity Hygroscopic
    Molecular Formula Cu(ClO4)2·6H2O
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    Odor Odorless
    MDL Number MFCD00149666
    Merck Index 14,2647
    Quantity 10g
    TSCA TSCA
    UN Number UN1481
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.23g/mL
    Refractive Index 1.505
    CAS 10294-46-9
    Melting Point 82°C
    Health Hazard 1 Danger
    Chemical Name or Material Copper (II) perchlorate hexahydrate
    Assay 99.999%, (metals basis)

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit