Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) picolinate, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
14050-01-2
C12H8CuN2O4
307.75
GNWVUSJJFONIKF-UHFFFAOYSA-L
copper ii picolinate,unii-96e596pypp,copper picolinate,copper, bis picolinato,copper picolinate anhydrous,copper, bis 2-pyridinecarboxylato-kappan1,kappao2,copper 2+ ion dipicolinate,copper,bis 2-pyridinecarboxylato-kn1,ko2-, hydrate 1:2
14963587
copper;pyridine-2-carboxylate
C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cu+2]
14050-01-2
C12H8CuN2O4
307.75
GNWVUSJJFONIKF-UHFFFAOYSA-L
copper ii picolinate,unii-96e596pypp,copper picolinate,copper, bis picolinato,copper picolinate anhydrous,copper, bis 2-pyridinecarboxylato-kappan1,kappao2,copper 2+ ion dipicolinate,copper,bis 2-pyridinecarboxylato-kn1,ko2-, hydrate 1:2
14963587
copper;pyridine-2-carboxylate
C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cu+2]
Assay Percent Range 97.5% min. (Iodometry)
Formula Weight 307.75
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 0.02
Quantity 25g
Chemical Name or Material Copper(II) picolinate

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Very toxic to aquatic life with long lasting effects.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning