Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) selenite hydrate, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4022118

Quantity 50g

 View more versions of this product

Catalog No. AA4022118


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10214-40-1
97%
190.50 (Anhydrous)
Copper(II) selenite hydrate
Crystalline Powder
CuO3Se·xH2O
MFCD00010973

Specifications

10214-40-1
97%
190.50 (Anhydrous)
MFCD00010973
50g
Crystalline Powder
CuO3Se·xH2O
Blue crystalline powder
Blue
Copper(II) selenite hydrate

Safety and Handling

SDS & Safety Documents

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packing Group: I

EINECSNumber : 233-526-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit