Copper(II) selenite hydrate, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar 4022106

Quantity 5g

 View more versions of this product

Catalog No. AA4022106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10214-40-1
    Physical Form Crystalline Powder
    Assay Percent Range 97%
    Molecular Formula CuO3Se·xH2O
    Formula Weight 190.50 (anhydrous)
    MDL Number MFCD00010973
    Chemical Name or Material Copper(II) selenite hydrate

    Specifications

    Formula Weight 190.50 (anhydrous)
    Quantity 5g
    Color Blue
    DOT Information DOT Class: 6.1, Packing Group: I
    EINECS Number 233-526-6
    Physical Form Crystalline Powder
    Molecular Formula CuO3Se·xH2O
    Appearance Blue crystalline powder
    MDL Number MFCD00010973
    CAS 10214-40-1
    Assay Percent Range 97%
    Health Hazard 1 H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Copper(II) selenite hydrate

    H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit