Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate, 98+%, for analysis, anhydrous, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
7758-98-7
CuO4S
159.6
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper sulfate,copper ii sulfate,cupric sulfate anhydrous,copper sulphate,copper 2+ sulfate,blue stone,copper monosulfate,copper ii sulfate, anhydrous,hylinec,trinagle
24462
CHEBI:23414
copper;sulfate
[O-]S(=O)(=O)[O-].[Cu+2]
7758-98-7
CuO4S
159.6
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper sulfate,copper ii sulfate,cupric sulfate anhydrous,copper sulphate,copper 2+ sulfate,blue stone,copper monosulfate,copper ii sulfate, anhydrous,hylinec,trinagle
24462
CHEBI:23414
copper;sulfate
[O-]S(=O)(=O)[O-].[Cu+2]
For analysis
Assay Percent Range 98% min. (Iodometry)
Linear Formula CuSO4
Fieser 01,164; 02,89; 05,162; 06,141; 08,125; 10,107; 16,283
Merck Index 15,2643
Solubility Information Solubility in water: 203g/L (20°C).
Formula Weight 159.6
Physical Form Fine Powder
Percent Purity 98+%
Grade Analysis
Calcium (Ca) 20ppm max.
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Loss on Drying 1% (300°C) max.
Magnesium (Mg) 50ppm max.
Nickel (Ni) 50ppm max.
Packaging Glass bottle
Solubility Solubility in water: 203g/L (20°C).
Total Nitrogen 0.01% max.
Zinc (Zn) 20ppm max.
Color Gray to Green
Melting Point 200.0°C
Quantity 100g
Chemical Name or Material Copper(II) sulfate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y

GHS Signal Word: Warning

EINECSNumber : 231-847-6