Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Copper(Ii) Sulfate Hydrate, 98%, Honeywell™ Fluka™

Click to view available options
Quantity:
25g
1kg
Packaging:
Poly Bottle
23254-43-5
CuH2O5S
177.617
MFCD00149682
CYKLGTUKGYURDP-UHFFFAOYSA-L
copper sulfate monohydrate,cupric sulfate monohydrate,copper sulfate, monohydrate,copper sulfate hydrate,caswell no. 257,epa pesticide chemical code 024402,copper ii sulfate hydrate,sulfuric acid copper 2+ salt 1:1 , monohydrate,copper sulphate hydrate,cuso4.h2o
61499
copper;sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Cu+2]
23254-43-5
CuH2O5S
177.617
MFCD00149682
CYKLGTUKGYURDP-UHFFFAOYSA-L
copper sulfate monohydrate,cupric sulfate monohydrate,copper sulfate, monohydrate,copper sulfate hydrate,caswell no. 257,epa pesticide chemical code 024402,copper ii sulfate hydrate,sulfuric acid copper 2+ salt 1:1 , monohydrate,copper sulphate hydrate,cuso4.h2o
61499
copper;sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Cu+2]

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula CuSO4 · xH2O
UN Number UN3077
Appearance Crystals/Powder
Formula Weight 159.61g/mol (anhydrous basis)
Percent Purity 98%
Density 200°C
Packaging Poly Bottle
Vapor Pressure 7.3mmHg (25°C)
Color Blue or White
Melting Point 3603g/cu.cm
Quantity 1kg
Chemical Name or Material Copper(Ii) Sulfate Hydrate
Show More Show Less

P273-P305 + P351 + P338-P501

H302-H315-H319-H410

DOTInformation : 9/PGIII

EINECSNumber : 231-847-6