Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper (II) Sulfate Pentahydrate, 10% w/v, Reagents

Catalog No. 01335665
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Catalog No. Quantity
01-335-665 1 L
01-335-666 10 L
01-335-664 4 L
01-335-663 500 mL
4 options

Catalog No. 01-335-665

Supplier: Reagents Holdings Llc CC1025001A

Only null left
Add to Cart
Add to Cart

Copper (II) Sulfate Pentahydrate, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7758-99-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Copper(II) Sulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Blue
Physical Form Liquid
Quantity 1 L
Percent Purity 10% w/v
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.064
Chemical Name or Material Copper (II) Sulfate Pentahydrate, 10% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.