Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate pentahydrate, 98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic Bottle
7758-99-8
CuO45H2O
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
7758-99-8
CuO45H2O
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
Additional Information contains an anticaking agent: < 1%
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula CuSO4·5H2O
Fieser 01,164; 02,89; 05,162; 06,141; 14,267
Merck Index 15, 2643
Arsenic (As) 10ppm max.
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol,slightly soluble in alcohol and glycerol,insoluble in ethanol
Formula Weight 249.68
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Grade Extra Pure
Iron (Fe) 200ppm max.
Lead (Pb) 125ppm max.
Nickel (Ni) 100ppm max.
Packaging Plastic Bottle
Solubility Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in alcohol and glycerol, insoluble in ethanol
Color Blue
Melting Point 110.0°C
Quantity 1kg
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning