Learn More
Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
Description
Copper(II) sulfide is used in antifouling paints, mixed catalysts and to develop aniline black dyes in textile printing. Further, it is used as a varnish pigment and an oxidizing agent. In addition, it is used as a component of solid state electrodes and thin-film solar cell with cadmium sulfide. It is a moderate conductor of electricity and used in photovoltaics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.8% (Metal basis) |
Solubility Information | Soluble in nitric acid,ammonium hydroxide,potassium cyanide. Insoluble in water,ethanol,hydrochloric acid ,dilute acids,sulfuric acid,alkali cyanides,aqueous ammonia and alkaline solutions. |
IUPAC Name | sulfanylidenecopper |
Formula Weight | 95.6 |
Odor | Odorless |
Refractive Index | 1.45 |
Show More |
Safety and Handling
EINECSNumber | 215-271-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.