Alfa Aesar™ Copper(II) sulfide, ultra dry, 99.98% (metals basis)

Manufacturer: Alfa Aesar™ 1430018

Quantity 50g

 View more versions of this product

Catalog No. AA1430018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-40-4
    Formula Weight 95.6
    MDL Number MFCD00016066
    Merck Index 14,2965
    Chemical Name or Material Copper(II) sulfide, Ultra dry

    Specifications

    Formula Weight 95.6
    Packaging Ampule
    Odor Odorless
    Sensitivity Air & Moisture Sensitive
    Melting Point 220°C (dec.)
    Physical Form Powder
    MDL Number MFCD00016066
    Merck Index 14,2965
    Quantity 50g
    CAS 1317-40-4
    Density 3.9g/mL to 4.6g/mL
    Refractive Index 1.45
    Storage Note 1
    • Ampouled under argon
    TSCA TSCA
    Chemical Name or Material Copper(II) sulfide, Ultra dry
    Assay 99.98%, (metals basis)

    • Ampouled under argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit