Copper(II) trifluoroacetate hydrate, Alfa Aesar™

Manufacturer: Alfa Aesar 4462409

Quantity 10g

 View more versions of this product

Catalog No. AA4462409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123333-88-0
    Physical Form Crystalline
    Molecular Formula Cu(OOCCF3)2·xH2O
    Formula Weight 289.57 (Anhydrous)
    MDL Number MFCD00013200
    Chemical Name or Material Copper(II) trifluoroacetate hydrate

    Specifications

    Formula Weight 289.57 (Anhydrous)
    Quantity 10g
    Physical Form Crystalline
    Molecular Formula Cu(OOCCF3)2·xH2O
    MDL Number MFCD00013200
    CAS 123333-88-0
    Chemical Name or Material Copper(II) trifluoroacetate hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit