Learn More
Copper(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Description
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | (decomposition) |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 4028198 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 361.68 |
Percent Purity | 98% |
Chemical Name or Material | Copper(II) trifluoromethanesulfonate |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 252-300-8 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.