Alfa Aesar™ Copper Manganese Nickel wire, 1.0mm (0.04 in.) dia.

Manufacturer: Alfa Aesar™ 43191G9

Quantity 25m

 View more versions of this product

Catalog No. AA43191G9

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01866121
    Chemical Name or Material Copper Manganese Nickel wire, 1.0mm (0.04 in.) dia.

    Specifications

    TSCA TSCA
    Physical Form Wire
    MDL Number MFCD01866121
    Avg. Mol. Wt. or Mol. Wt. Range Cu:Mn:Ni; 84:12:4 wt%
    Weight ≈6.6g/m
    Quantity 25m
    Chemical Name or Material Copper Manganese Nickel wire, 1.0mm (0.04 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit