Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Metal, 0.02 in. Diameter, Wire, Spectrum™

Click to view available options
Quantity:
100 g
500 g
Packaging:
Fiber Tube
7440-50-8
7440-50-8
Quantity 100 g
CAS Max % 1
Packaging Fiber Tube
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.