Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Metal, Granular, Approx. 98%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 lb.
Packaging:
Poly Bottle
Poly Pail
7440-50-8
7440-50-8
Quantity 125 g
CAS Max % 1
Packaging Poly Bottle
Percent Purity Approx. 98%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.