Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Metal, Shot, 99.9%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
7440-50-8
7440-50-8
Quantity 500 g
CAS Max % 1
Packaging Poly Bottle
Percent Purity 0.999
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.